Incinerating dishes
Leverancier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APG210X170X300 APG220X170X100 APG280X210X100 APG350X230X58 APG350X240X40 APG420X200X50
HALDAPG210X170X300EA 1900 EUR
HALDAPG210X170X300 HALDAPG220X170X100 HALDAPG280X210X100 HALDAPG350X230X58 HALDAPG350X240X40 HALDAPG420X200X50
Incinerating dishes
Schuitjes Verbrandingsschuitjes
Alsint 99,7.
  • Curved suvertical surfaces for easy removal of sample
  • Good thermal shock resistance due to high thermal conductivity
  • Resistant to chemical attack, e.g. hydrogen and other reducing gases