Laboratory bottles, narrow neck, with screw cap
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632414321950 632414321956 632414321966 632414321968 632414345100_U 632414345500_U 632414345250_U 632414345956_U 632414345950 632414345966 632414645968 632414345100 632414345500 632414345250 632414345956 632414321100 632414321500 632414321940 632414321250 632414501050 632414345940
KAVA632414321950EA 44.7 EUR
KAVA632414321950 KAVA632414321956 KAVA632414321966 KAVA632414321968 KAVA632414345100_U KAVA632414345500_U KAVA632414345250_U KAVA632414345956_U KAVA632414345950 KAVA632414345966 KAVA632414645968 KAVA632414345100 KAVA632414345500 KAVA632414345250 KAVA632414345956 KAVA632414321100 KAVA632414321500 KAVA632414321940 KAVA632414321250 KAVA632414501050 KAVA632414345940
Laboratory bottles, narrow neck, with screw cap
Flessen Glazen flessen
Borosilicate glass 3,3, clear or brown.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round, with graduation, DIN thread GL 45, pouring ring and blue screw cap.
Certificaten: In compliance with DIN ISO 4796 - 1 standards