Fused silica tubing
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091-21050 19091-60620E 160-1010-1 160-1010-10 160-1010-5 160-2200-10 160-2200-5 160-2205-1 160-2205-10 160-2205-5 160-2250-10 160-2250-5 160-2255-1 160-2255-10 160-2255-30 160-2255-5 160-2320-10 160-2320-5 160-2325-1 160-2325-10 160-2325-30 160-2325-5 160-2455-1 160-2455-10 160-2455-5 160-2530-10 160-2530-5 160-2535-1 160-2535-10 160-2535-30 160-2535-5 160-2610-10 160-2610-5 160-2615-1 160-2615-10 160-2615-5 160-2634-10 160-2634-5 160-2635-1 160-2635-10 160-2635-5 160-2644-10 160-2650-10 160-2655-1 160-2655-10 160-2655-5 160-2815-5 160-2825-5 160-2845-10 160-2845-5 160-2855-10 160-2855-5 160-2865-10 160-2865-5 160-4535-5 CP8016 160-2625-1 160-2625-10 160-2625-5 CP68186 CP68187 CP8003 CP801206 CP801505 CP801510 CP801805 CP801806 CP801810 CP802505 CP802510 CP802530 CP803205 CP803210 CP805305 CP805306 CP805310 CP805330
HEWL19091-21050EA 436 EUR
HEWL19091-21050 HEWL19091-60620E HEWL160-1010-1 HEWL160-1010-10 HEWL160-1010-5 HEWL160-2200-10 HEWL160-2200-5 HEWL160-2205-1 HEWL160-2205-10 HEWL160-2205-5 HEWL160-2250-10 HEWL160-2250-5 HEWL160-2255-1 HEWL160-2255-10 HEWL160-2255-30 HEWL160-2255-5 HEWL160-2320-10 HEWL160-2320-5 HEWL160-2325-1 HEWL160-2325-10 HEWL160-2325-30 HEWL160-2325-5 HEWL160-2455-1 HEWL160-2455-10 HEWL160-2455-5 HEWL160-2530-10 HEWL160-2530-5 HEWL160-2535-1 HEWL160-2535-10 HEWL160-2535-30 HEWL160-2535-5 HEWL160-2610-10 HEWL160-2610-5 HEWL160-2615-1 HEWL160-2615-10 HEWL160-2615-5 HEWL160-2634-10 HEWL160-2634-5 HEWL160-2635-1 HEWL160-2635-10 HEWL160-2635-5 HEWL160-2644-10 HEWL160-2650-10 HEWL160-2655-1 HEWL160-2655-10 HEWL160-2655-5 HEWL160-2815-5 HEWL160-2825-5 HEWL160-2845-10 HEWL160-2845-5 HEWL160-2855-10 HEWL160-2855-5 HEWL160-2865-10 HEWL160-2865-5 HEWL160-4535-5 VARICP8016 HEWL160-2625-1 HEWL160-2625-10 HEWL160-2625-5 VARICP68186 VARICP68187 VARICP8003 HEWLCP801206 HEWLCP801505 HEWLCP801510 HEWLCP801805 HEWLCP801806 HEWLCP801810 HEWLCP802505 HEWLCP802510 HEWLCP802530 HEWLCP803205 HEWLCP803210 HEWLCP805305 HEWLCP805306 HEWLCP805310 HEWLCP805330
Fused silica tubing
Slangen Chromatography Tubing
Deactivated or undeactivated fused silica tubing.
Deactivated tubing can be used as retention gaps, guard columns or transfer lines. The standard deactivation process is a phenyl methyl deactivation. - the preferred choice for most applications due to its inertness and robustness.
Undeactivated tubing or bar fused silica is commonly used for capillary electrophoresis. It can also be used for transfer lines and other applications where inertness is not critical.