Specimen jars with knobbed drop-on cover
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632416130808 632416131212 632416131515 632416132020
216-3351EA 109 EUR
216-3351 216-3353 216-3354 216-3355
Specimen jars with knobbed drop-on cover
Potjes Specimen Potten
Made of glass with optical purity.
  • Sturdy
  • Excellent chemical resistance
  • Does not disfigure the view of content
Specimen jars with base and drop-on cover are suitable for use in biology and medicine for specimen storage.