Dropping bottles, glass, with ground-in pipette stopper
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632425020050 632425020100 632425020250 632425023050 632425023100 632425023250
215-6246EA 39.1 EUR
215-6246 215-6247 215-6248 215-6265 215-6266 215-6267
Dropping bottles, glass, with ground-in pipette stopper
Flessen Druppelflessen
Glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette
Amber to protect light-sensitive substances.