Capillary columns, OPTIMA® 1301
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
726771.25 726771.30 726771.50 726771.60 726777.25 726777.30 726777.60 726780.30 726780.50 726780.60 726783.25
MANA726771.25EA 632 EUR
MANA726771.25 MANA726771.30 MANA726771.50 MANA726771.60 MANA726777.25 MANA726777.30 MANA726777.60 MANA726780.30 MANA726780.50 MANA726780.60 MANA726783.25
Capillary columns, OPTIMA® 1301
Kolommen voor chromatografie
Medium polar GC phase.
  • Ideal for pesticide analyses
  • Maximum temperature for isothermal operation 300 °C
  • Max. temperature for short isotherms in a temperature program 320 °C
6% cyanopropyl-phenyl – 94% dimethylpolysiloxane phase.
USP code: G43.