Filtration crucibles
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3660/02M 3660/04M 3660/06M 3660/08M 3680/02M 3680/04M 3680/06M 3680/08M 3680/10M 3650/02M 3660/10M 3650/08M 3650/04M
459-0242EA 117 EUR
459-0242 459-0234 459-0235 459-0236 459-0244 459-0245 459-0246 459-0247 459-0248 459-0240 BIBB3660/10M 459-0241 BIBB3650/04M
Filtration crucibles
Kroezen
These filtration crucibles are made from Pyrex® borosilicate glass.
  • With integral glass sintered disc
  • Ideal for drying precipitates to constant mass at 110 °C
  • Suitable for ignition work up to 450 °C