Polyethylene stoppers
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TRF591 TRF592 TRF593 TRF595 TRF596 TRF597 TRF598
217-0716EA 30.8 EUR
217-0716 217-0717 217-0718 AZLOTRF595 AZLOTRF596 AZLOTRF597 AZLOTRF598
Polyethylene stoppers
Stoppen PE/LDPE/HDPE stoppen
These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets.
Large head provides a good grip and gives protection if glassware is accidently knocked over.
Verpakking: 10/package