Joint clips for spherical joints, metal, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JC13 JC19
201-0761EA 84.7 EUR
201-0761 201-0565
Joint clips for spherical joints, metal, Quickfit®
Klemmen NS-verbindingsklemmen
Metal, corrosion resistant nickel finish. Clips in 'fork' type design for retaining glassware having Quickfit® spherical joints.
  • Spring loaded to ensure a positive pressure is applied to both surfaces of the joint
  • Suitable for high temperature work