PYREX® Bottles, Media-lab, Clear, With Cap and Pouring Ring
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1516/04D 1516/06D 1516/08D 1516/01D 1516/02D 1516/10D 1516/12D 1516/16D 1516/18D 1516/22D 1516/20D 1516/05D 1516/14D
215-3123EA 107 EUR
215-3123 215-3124 215-3127 215-3110 215-3122 215-3128 215-3537 215-3538 215-3539 BIBB1516/22D BIBB1516/20D 215-3540 BIBB1516/14D
PYREX® Bottles, Media-lab, Clear, With Cap and Pouring Ring
Flessen Glazen flessen
Since this bottle was first introduced in the 1980’s, the versatile design has become the most commonly used laboratory bottle. Originally designed for the preparation and storage of culture media, this bottle is ideal for many applications.
  • Printed with Trace Code for downloadable batch certificate
  • Ideal for mixing and storage of culture medium
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • For use up to 140 °C
Bottles are manufactured from temperature-resistant and chemically-resistant borosilicate glass. All bottles are supplied complete with polypropylene cap and drip-free pouring ring.
The 25 ml size bottle features a glass lip and does not require a pouring ring.
Certificaten:
Complies with ISO 4796.