Screw cap for reagent bottles
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4503/60 4513/60M 4503/13 4503/15 4503/18 4503/20 4503/22 4503/25 4503/30 4503/42 4513/13M 4513/15M 4513/18M 4513/20M 4513/22M 4513/25M 4513/30M 4513/42M
212-7482EA 103 EUR
212-7482 BIBB4513/60M BIBB4503/13 BIBB4503/15 BIBB4503/18 BIBB4503/20 BIBB4503/22 BIBB4503/25 215-3135 BIBB4503/42 BIBB4513/13M BIBB4513/15M BIBB4513/18M BIBB4513/20M BIBB4513/22M BIBB4513/25M 215-3137 BIBB4513/42M
Screw cap for reagent bottles
Sluitingen Flesdoppen
Phenolic cap with PTFE faced sealing disc
  • Fits PYREX® reagent bottles
  • Excellent chemical resistance
  • Autoclavable up to 121 °C