Flat flange lids with 5 necks
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
296-00 296-02 296-03
BIBB296-00EA 174 EUR
BIBB296-00 BIBB296-02 BIBB296-03
Flat flange lids with 5 necks
Deksels Deksels voor reactievaten
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For use with SVL® reaction vessels. Available without, with two or five necks. Necks with GL thread.