Centrifuge tubes, glass
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1660/02M 1660/06M 1660/08M
525-0520EA 325 EUR
525-0520 BIBB1660/06M BIBB1660/08M
Centrifuge tubes, glass
Buisjes Centrifugebuisjes
PYREX®, borosilicate glass, cylindrical with round bottom.
  • Uniform glass distribution in wall and base for improved strength
  • Excellent chemical and thermal resistance
  • Max. RCF 3000×g
Robust universal centrifuge tubes for many uses.