Standard conical ground joints cones, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CNB40UB CNB10UB CNB34UB CNB14UB CNB19UB CNB24UB CNB29UB CNB45UB CNB7UB
201-8768EA 70.7 EUR
201-8768 201-8197 201-8198 201-0072 201-0073 201-0074 201-0075 BIBBCNB45UB BIBBCNB7UB
Standard conical ground joints cones, Quickfit®
Adapters Adapters met NS-verbindingen
PYREX® borosilicate glass.
  • Resistance to chemical corrosion and thermal shock
  • Manufactured to an accuracy which surpasses the requirements of ISO 383
  • Avoids the problem of the badge charring at high temperatures which can give an untidy appearance
Unbadged ground glass cones free from any ceramic markings.
Total length: 100 mm