Standard conical ground joints sockets, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SRB10UB SRB40UB SRB34UB SRB14UB SRB19UB SRB24UB SRB29UB SRB45UB
201-8194EA 91.8 EUR
201-8194 BIBBSRB40UB 201-8195 201-0076 201-0077 201-0078 201-0079 BIBBSRB45UB
Standard conical ground joints sockets, Quickfit®
Adapters Adapters met NS-verbindingen
PYREX® borosilicate glass.
  • Accurate taper and fine surface finish reduce possibility of joint 'seizure' or leakage
  • Manufactured to an accuracy which surpasses the requirements of ISO 383
  • Avoids the problem of the badge charring at high temperatures which can give an untidy appearance
Unbadged ground glass sockets free from any ceramic markings.
Total length: 100 mm