2-Formylfenylboronzuur (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Formylbenzeneboronic acid, (2-Formylphenyl)metaboric acid, 2-Boronobenzaldehyde, (2-Formylphenyl)boronic acid, Dihydroxy(2-formylphenyl)borane
B1873-1G
B1873-5G
TCIAB1873-1GEA
25.3
EUR
TCIAB1873-1G
TCIAB1873-5G
2-Formylfenylboronzuur (contains varying amounts of Anhydride)
2-Formylfenylboronzuur
Formule:
C₇H₇BO₃ MW: 149.94 g/mol |
MDL Nummer:
MFCD00151822 CAS nummer: 40138-16-7 |
|
Specification Test Results
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...