2-Nitro-p-xylene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D1105-25G D1105-500G
TCIAD1105-25GEA 50.6 EUR
TCIAD1105-25G TCIAD1105-500G
2-Nitro-p-xylene ≥99.0% (by GC)
2-Nitro-p-xylene
Formule: C₈H₉NO₂
Kookpunt: 104°C
Smeltpunt: –25 °C
Dichtheid: 1,132 g/cm³ (20 °C)
Vlampunt: 104°C
MDL Nummer: MFCD00024284
CAS nummer: 89-58-7
EINECS: 201-920-7
UN: 1665
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b28e4543958694df2577aa742dec7892f1f51d&vid=c3b28e4543958694df2577aa742dec7892f1f51d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 99.0 %