Kresol
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C0412-25G
C0412-500G
TCIAC0412-25GEA
17.6
EUR
TCIAC0412-25G
TCIAC0412-500G
Kresol
Kresol
Formule:
C₇H₈O Kookpunt: 192°C Dichtheid: 1,03 g/cm³ (20 °C) Vlampunt: 81°C |
MDL Nummer:
MFCD00151099 CAS nummer: 1319-77-3 EINECS: 215-293-2 UN: 2022 ADR: 6.1,II Merck Index: 14,02579 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
m-Cresol | 50.0 to 60.0 % |
p-Cresol | 25.0 to 35.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...