VWR® AVS TITRINORM, Dysprosium Standard Solution, 1000 mg/L Dy in dil. Nitric Acid AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86674.180EA
24.1
EUR
86674.180
86674.260
VWR® AVS TITRINORM, Dysprosium Standard Solution, 1000 mg/L Dy in dil. Nitric Acid AVS TITRINORM Standard for AAS
Standaarden
Kationische standaarden
Dysprosium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%