4,4-Dimethoxybutanone ≥90.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1157-25ML D1157-500ML D1157-100ML
TCIAD1157-25MLEA 17.6 EUR
TCIAD1157-25ML TCIAD1157-500ML TCIAD1157-100ML
4,4-Dimethoxybutanone ≥90.0% (by GC)
4,4-Dimethoxybutanone
Formule: C₆H₁₂O₃
Kookpunt: 178 °C (1013 hPa)
Smeltpunt: –82 °C
Dichtheid: 0,9451 g/cm³ (20 °C)
Vlampunt: 58°C
MDL Nummer: MFCD00008789
CAS nummer: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3a7e84dbaf0b99cd39c7f48ca00fbb62588416&vid=dc3a7e84dbaf0b99cd39c7f48ca00fbb62588416&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %