3-Nitrobenzyl alcohol ≥99.0% (by GC) matrix for FABMS and liquid SIMS

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (3-Nitrophenyl)methanol

S0378-1G S0378-10G
TCIAS0378-1GEA 19.8 EUR
TCIAS0378-1G TCIAS0378-10G
3-Nitrobenzyl alcohol ≥99.0% (by GC) matrix for FABMS and liquid SIMS
3-Nitrobenzyl alcohol
Formule: C₇H₇NO₃
Kookpunt: 317 °C (1013 hPa)
Smeltpunt: 26…27 °C
Dichtheid: 1,3 g/cm³ (20 °C)
MDL Nummer: MFCD00007273
CAS nummer: 619-25-0
EINECS: 210-588-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3af97e9e63692fca90a8a4aa05f90bfdd9be93&vid=a8d4b34f6fca5018cf73e6ade894c6931836dd47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR