Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Calcium sulphate (anhydrous) anhydrite, Gypsum
89750.TD 89750.XA
89750.TDEA 38.3 EUR
89750.TD 89750.XA
Calcium sulphate, Drierite™ 3 mm (8 mesh) for drying
Calcium sulphate
CAS No.: 7778-18-9 Physical Form: Granular -8 Mesh Sensitivity: Hygroscopic Appearance: White Odour: Odourless Merck Reference: 14,1706 MDL No.: MFCD00010912
Formule: CaO₄S
MW: 136.14 g/mol
Smeltpunt: 1350
Dichtheid: 2.964 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010912
CAS nummer: 7778-18-9
EINECS: 231-900-3
Merck Index: 14,1706