Phloroglucinol, watervrij ≥99.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P0249-250G P0249-25G
TCIAP0249-250GEA 395 EUR
TCIAP0249-250G TCIAP0249-25G
Phloroglucinol, watervrij ≥99.0% (by HPLC)
Phloroglucinol
Formule: C₆H₆O₃
Kookpunt: 400 °C (1013 hPa)
Smeltpunt: 217…220 °C
Dichtheid: 1,46 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002286
CAS nummer: 108-73-6
EINECS: 203-611-2
Merck Index: 14,07328
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2d2aa4ecaba2203120d1695ac7cc3a1bb0956fe&vid=011d775ab3450f2bca1d3179630757663d8016b2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 215.0 to 220.0 deg-C
Solubility in EtOH almost transparency