1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 1H,1H,2H,2H-Perfluorooctyl iodide, 1-Iodo-3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctane
T2074-25G T2074-5G
TCIAT2074-25GEA 86.9 EUR
TCIAT2074-25G TCIAT2074-5G
1H,1H,2H,2H-Perfluoro-1-iodooctane ≥97.0% (by GC)
1H,1H,2H,2H-Perfluoro-1-iodooctane
Formule: C₈H₄F₁₃I
Kookpunt: 180°C
Dichtheid: 1,91 g/cm³ (25 °C)
Vlampunt: 150°C
MDL Nummer: MFCD00039410
CAS nummer: 2043-57-4
EINECS: 218-056-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=993951b1984d758598136a387c75c2cd1d650bd7&vid=993951b1984d758598136a387c75c2cd1d650bd7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %