Diethyl sulphide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

E0176-25ML E0176-100ML
TCIAE0176-25MLEA 45.1 EUR
TCIAE0176-25ML TCIAE0176-100ML
Diethyl sulphide ≥98.0% (by GC)
Diethyl sulphide
Formule: C₄H₁₀S
MW: 90.19 g/mol
Kookpunt: 92°C
Smeltpunt: -104
Vlampunt: –10°C
MDL Nummer: MFCD00009270
CAS nummer: 352-93-2
EINECS: 206-526-9
UN: 2375
Merck Index: 14,03854
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65ef21662c7b2da6d8c12f2e4474a9171baaba48&vid=65ef21662c7b2da6d8c12f2e4474a9171baaba48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR