N,N-Dimethylformamide di-tert-butyl acetal ≥98.0% (by total nitrogen basis) for esterification

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D1303-25ML D1303-5ML
TCIAD1303-25MLEA 284 EUR
TCIAD1303-25ML TCIAD1303-5ML
N,N-Dimethylformamide di-tert-butyl acetal ≥98.0% (by total nitrogen basis) for esterification
N,N-Dimethylformamide di-tert-butyl acetal
Formule: C₁₁H₂₅NO₂
Kookpunt: 57°C
Dichtheid: 0,85 g/cm³ (20 °C)
Vlampunt: 33°C
MDL Nummer: MFCD00015002
CAS nummer: 36805-97-7
EINECS: 253-222-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ff18009e98f842ee2c06555166a532463408952&vid=f45857cedac8441de1e2c3c05f6ea742ec667397&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (with Total Nitrogen) min. 98.0 %
Effect test of GC derivalization Effective as derivatization agent Myristic Acid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR