Benzene-1,3,5-tricarbonyl trichloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1,3,5-Benzenetricarboxylic acid chloride, 1,3,5-Benzenetricarbonyl chloride, Trimesoylchloride, Trimesoyl chloride
T1262-25G T1262-250G
TCIAT1262-25GEA 64.9 EUR
TCIAT1262-25G TCIAT1262-250G
Benzene-1,3,5-tricarbonyl trichloride ≥98.0% (by GC, titration analysis)
Benzene-1,3,5-tricarbonyl trichloride
Formule: C₉H₃Cl₃O₃
MW: 265.48 g/mol
Kookpunt: 181°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000679
CAS nummer: 4422-95-1
EINECS: 224-594-8
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9941df094b998d4d516c37bd5e3b4f05e0c9d036&vid=9941df094b998d4d516c37bd5e3b4f05e0c9d036&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Toluene almost transparency