VWR® AVS TITRINORM, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86669.180EA 38.3 EUR
86669.180 86669.260
VWR® AVS TITRINORM, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Cerium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%