VWR®, Gallium standard solution, 1,000 mg/L Ga in dilute nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86678.180EA
74
EUR
86678.180
86678.260
VWR®, Gallium standard solution, 1,000 mg/L Ga in dilute nitric acid AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Gallium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%