VWR® AVS TITRINORM, Indium Standard Solution, 1000 mg/L In in dilute Nitric Acid AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86684.180EA 44.1 EUR
86684.180 86684.260
VWR® AVS TITRINORM, Indium Standard Solution, 1000 mg/L In in dilute Nitric Acid AVS TITRINORM Standard for AAS
Standaarden Kationische standaarden Indium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
Limited Time Sale
Select items below are currently on sale
Order Now
Specificaties
Environmentally Preferable