VWR®, Platinum standard solution, 1,000 mg/L Pt in 10% hydrochloric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86701.260EA
319
EUR
86701.260
86701.180
VWR®, Platinum standard solution, 1,000 mg/L Pt in 10% hydrochloric acid AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Platina standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%