Dihexyl ether ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
H0138-25ML H0138-500ML
TCIAH0138-25MLEA 35.2 EUR
TCIAH0138-25ML TCIAH0138-500ML
Dihexyl ether ≥98.0% (by GC)
Dihexyl ether
Formule: C₁₂H₂₆O
MW: 186.34 g/mol
Kookpunt: 226°C
Vlampunt: 77°C
MDL Nummer: MFCD00009525
CAS nummer: 112-58-3
EINECS: 203-987-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc35ee2bdf89c6551fe2075abd898f31ec850f20&vid=cc35ee2bdf89c6551fe2075abd898f31ec850f20&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %