VWR® AVS TITRINORM, Hafnium Standard Solution, 1000 mg/L Hf in Dilute Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86681.180EA
77.7
EUR
86681.180
86681.260
VWR® AVS TITRINORM, Hafnium Standard Solution, 1000 mg/L Hf in Dilute Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
Standaarden
Kationische standaarden
Hafnium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%