VWR® AVS TITRINORM, Iridium Standard Solution, 1,000 mg/L Ir in 10% Hydrochloric Acid AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86685.180EA 139.3 EUR
86685.180 86685.260
VWR® AVS TITRINORM, Iridium Standard Solution, 1,000 mg/L Ir in 10% Hydrochloric Acid AVS TITRINORM Standard for AAS
Standaarden Kationische standaarden Iridium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%