VWR®, Phosphorus standard solution, 1,000 mg/L P in water AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86698.180EA
25.8
EUR
86698.180
86698.260
VWR®, Phosphorus standard solution, 1,000 mg/L P in water AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Fosfor standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%