VWR® AVS TITRINORM, Scandium Standard Solution, 1000 mg/L Sc in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86708.180EA 74.2 EUR
86708.180 86708.260
VWR® AVS TITRINORM, Scandium Standard Solution, 1000 mg/L Sc in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Scandium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%