Butylmethacrylaat ≥99.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Methacrylic acid butyl ester
Stabilisatie: Stabilised with hydroquinone (HQ)
M0081-25ML M0081-500ML
TCIAM0081-25MLEA 17.6 EUR
TCIAM0081-25ML TCIAM0081-500ML
Butylmethacrylaat ≥99.0% (by GC) stabilised
Butylmethacrylaat
Formule: C₈H₁₄O₂
MW: 142.2 g/mol
Kookpunt: 164°C
Smeltpunt: –25°C
Vlampunt: 51°C
MDL Nummer: MFCD00009444
CAS nummer: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2305e62130b6fcb7333e0b1315debb3dd53ee8c&vid=d2305e62130b6fcb7333e0b1315debb3dd53ee8c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %