Bicine ≥99.0% (by titrimetric analysis) good's buffer component for biological research
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
B0484-25G
B0484-500G
B0484-100G
TCIAB0484-25GEA
28.6
EUR
TCIAB0484-25G
TCIAB0484-500G
TCIAB0484-100G
Bicine ≥99.0% (by titrimetric analysis) good's buffer component for biological research
Bicine
Formule:
C₆H₁₃NO₄ MW: 163.17 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004295 CAS nummer: 150-25-4 EINECS: 205-755-1 Merck Index: 14,01201 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Absorbance | max. 0.15 (1mol/L, H2O, 250nm) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...