1-Iodoperfluorbutane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Perfluorobutyl iodide, 1,1,1,2,2,3,3,4,4-Nonafluoro-4-iodobutane, 1-Iodononafluorobutane, Nonafluoro-1-iodobutane
N0499-25G N0499-500G N0499-100G
TCIAN0499-25GEA 39.6 EUR
TCIAN0499-25G TCIAN0499-500G TCIAN0499-100G
1-Iodoperfluorbutane ≥98.0% (by GC)
1-Iodoperfluorbutane
Formule: C₄F₉I
MW: 345.93 g/mol
Kookpunt: 67°C
Smeltpunt: -2
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001062
CAS nummer: 423-39-2
EINECS: 207-025-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43953aab63e8baf3604a1d7ac50ddf8967ffbe1c&vid=fbb35dae343a0056fec64c77248ffeb441e372e1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Red clear liquid
Purity (GC) min. 98.0 %