1-Iodohexane ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with copper chip

I0059-25G I0059-250G I0059-100G
TCIAI0059-25GEA 40.7 EUR
TCIAI0059-25G TCIAI0059-250G TCIAI0059-100G
1-Iodohexane ≥97.0% (by GC) stabilised
1-Iodohexane
Formule: C₆H₁₃I
Kookpunt: 180°C
Smeltpunt: –68 °C
Dichtheid: 1,44 g/cm³ (20 °C)
Vlampunt: 61°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001102
CAS nummer: 638-45-9
EINECS: 211-339-0
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03511666ca89d474e1c552cd9f6bc4480fb81678&vid=6d671b1c77cc9a684b4fa4434bb59c62d1a245cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR