Diiodomethane ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Stabilisatie: Stabilised with copper chip
D0610-25G D0610-500G D0610-100G
TCIAD0610-25GEA 50.6 EUR
TCIAD0610-25G TCIAD0610-500G TCIAD0610-100G
Diiodomethane ≥98.0% (by GC) stabilised
Diiodomethane
Formule: CH₂I₂
Kookpunt: 181°C
Smeltpunt: 6 °C
Dichtheid: 3,32537 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001079
CAS nummer: 75-11-6
EINECS: 200-841-5
UN: 1760
ADR: 8,III
Merck Index: 14,06066
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4caa43e8a665215f5024e1f94307bad4a5222e65&vid=cc6d17b4b6bf28d0a060f8de2f6ed17282eb530b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %