2-Iodoethanol ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with copper chip
I0391-25G
I0391-5G
TCIAI0391-25GEA
63.8
EUR
TCIAI0391-25G
TCIAI0391-5G
2-Iodoethanol ≥98.0% (by GC) stabilised
2-Iodoethanol
Formule:
C₂H₅IO Kookpunt: 85°C Dichtheid: 2,202 g/cm³ (20 °C) Vlampunt: 65°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00002830 CAS nummer: 624-76-0 EINECS: 210-861-6 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...