1-Iodo-2-methylpropane ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Isobutyliodide
Stabilisatie: Stabilised with copper chip
I0322-25ML I0322-250ML
TCIAI0322-25MLEA 60.5 EUR
TCIAI0322-25ML TCIAI0322-250ML
1-Iodo-2-methylpropane ≥97.0% (by GC) stabilised
1-Iodo-2-methylpropane
Formule: C₄H₉I
MW: 184.02 g/mol
Kookpunt: 121°C
Smeltpunt: -90.7
Vlampunt: 0°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001084
CAS nummer: 513-38-2
EINECS: 208-160-5
UN: 2391
ADR: 3 II
Merck Index: 14,05144
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e1ef391dc005438210cc6e0f42711a4ee6db63&vid=630c009a593566e9689b96d6e2a0aba765e77d6e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Brown clear liquid
Purity (GC) min. 97.0 %