3-Tolualdehyde ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisatie: Stabilised with hydroquinone (HQ)

T0258-25ML T0258-5ML
TCIAT0258-25MLEA 49.5 EUR
TCIAT0258-25ML TCIAT0258-5ML
3-Tolualdehyde ≥97.0% (by GC) stabilised
3-Tolualdehyde
Formule: C₈H₈O
Kookpunt: 200°C
Dichtheid: 1,0535 g/cm³ (20 °C)
Vlampunt: 78°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003374
CAS nummer: 620-23-5
EINECS: 210-632-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b5cde7e1ca4f4dc49cc60c7ca2d91cee533a2e&vid=f6b5cde7e1ca4f4dc49cc60c7ca2d91cee533a2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR