Iodocyclohexane ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with copper chip

I0056-10G I0056-100G
TCIAI0056-10GEA 38.5 EUR
TCIAI0056-10G TCIAI0056-100G
Iodocyclohexane ≥98.0% (by GC) stabilised
Iodocyclohexane
Formule: C₆H₁₁I
Kookpunt: 180°C
Dichtheid: 1,625 g/cm³ (20 °C)
Vlampunt: 72°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003826
CAS nummer: 626-62-0
EINECS: 210-957-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47219e3f7b60584b110b88338065a0f641539e52&vid=47219e3f7b60584b110b88338065a0f641539e52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR