Lithiumaluminiumhydride ≥95.0% (by titrimetric analysis), powder
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Aluminium lithium hydride, Lithium tetrahydridoaluminate
L0203-100G
L0203-25G
TCIAL0203-100GEA
378
EUR
TCIAL0203-100G
TCIAL0203-25G
Lithiumaluminiumhydride ≥95.0% (by titrimetric analysis), powder
Lithiumaluminiumhydride
Formule:
H₄AlLi MW: 37.95 g/mol Smeltpunt: 125 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011075 CAS nummer: 16853-85-3 EINECS: 240-877-9 UN: 1410 ADR: 4.3 I Merck Index: 14,00350 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (NBS Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...