VWR® AVS TITRINORM, Beryllium Standard Solution, 1000 mg/L Be in 2% Hydrochloric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86665.180EA 85.5 EUR
86665.180 86665.260
VWR® AVS TITRINORM, Beryllium Standard Solution, 1000 mg/L Be in 2% Hydrochloric Acid, Standard for AAS
Standaarden Kationische standaarden Beryllium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%