VWR® AVS TITRINORM, Palladium Standard Solution, 1000 mg/l Pd in 5% Hydrochloric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86700.180EA 98 EUR
86700.180 86700.260
VWR® AVS TITRINORM, Palladium Standard Solution, 1000 mg/l Pd in 5% Hydrochloric Acid, Standard for AAS
Standaarden Kationische standaarden Palladium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%