VWR®, Rhodium standard solution, 1,000 mg/l Rh in 5% hydrochloric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86704.180EA
201.6
EUR
86704.180
86704.260
VWR®, Rhodium standard solution, 1,000 mg/l Rh in 5% hydrochloric acid AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Rodium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%