VWR® AVS TITRINORM, Ruthenium Standard Solution, 1000 mg/L Ru in 5% Hydrochloric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86705.180EA
80.9
EUR
86705.180
86705.260
VWR® AVS TITRINORM, Ruthenium Standard Solution, 1000 mg/L Ru in 5% Hydrochloric Acid, Standard for AAS
Standaarden
Kationische standaarden
Ruthenium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%